Anti-EYA2 Rabbit Polyclonal Antibody

Supplier: ProSci
58-943
76070-064EA 623.94 USD
76070-064
Anti-EYA2 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000

Type: Primary
Antigen: EYA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 216-244
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR