BioPure Silver Nanospheres, 1 mg/ml, PEG Surface, nanoComposix
Supplier: nanoComposix
AGGB100-5M AGGB50-1M AGGB50-5M AGGB50-10M AGGB100-1M AGGB100-10M
103370-006EA 1130 USD
103370-006 103369-896 103370-008 103369-894 103369-898 103370-004
BioPure Silver Nanospheres, 1 mg/ml, PEG Surface, nanoComposix
Beads Microcarriers
Purified, concentrated, high quality silver nanoparticles. Sterile and endotoxin-free. PEG for neutral surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/mL, 9.27 mM, 1000 ppm, ~40 to 135 OD [depending on size])
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra-pure (Milli-Q) DI water
  • BioPure materials ideal for in-vitro and in-vivo experiments, nanotoxicology
  • PEG surface provides strong steric stability in salts and alcohols
Spherical silver nanoparticles, 1 mg/mL (high concentration)
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (domestic). Cold packaging included. Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.