Stigmasterol

Supplier: ADIPOGEN CORP MS

AG-CN2-0412-G005 AG-CN2-0412-G001
102988-178EA 221.54 USD
102988-178 102988-176
Stigmasterol
Stigmasterol

Anti-hypercholestrolemic compound. Anti-inflammatory and immune-modulating effects. Anticancer compound. Chemopreventive. Cytostatic. Cell growth inhibitor. Antimutagenic. Potent in vitro antagonist of FXR (farnesoid X receptor). DNA polymerase beta inhibitor. Potent antioxidant, hypoglycemic and thyroid inhibiting agent. Anti-osteoarthritic. Decreases the expression of matrix metalloproteinases. Neuroprotective. Is used as a precursor for synthetic progesterone and vitamin D3 and is an intermediate in the biosynthesis of androgens, estrogens and corticoids.

Formula: C₂₉H₄₈O
MDL Number: MFCD00003630
CAS Number: 83-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR