N-Glycolylneuraminic acid, solid

Supplier: TORONTO RESEARCH CHEMICALS MS

Synonyms: Neu5Gc

G65000010MG
100575-470EA 233.93 USD
100575-470
N-Glycolylneuraminic acid, solid
N-Glycolylneuraminic acid

Used for regulation of N-glycolylneuraminic acid biosynthesis in rat and mouse liver.

Formula: C₁₁H₁₉NO₁₀
MW: 325.27 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Freezer
MDL Number: MFCD00057551
CAS Number: 1113-83-3

Order Now

Specification Test Results

Color White to off-white
Form Solid
Solubility Water
Storage Store at -20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR