Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite

Supplier: TORONTO RESEARCH CHEMICALS MS

Synonyms: Bis(2-cyanoethyl) diisopropylphosphoramiditeBis(2-cyanoethoxy)(diisopropylamino)phosphineBis(2-cyanoethoxy)-N,N-diisopropylaminophosphine

B4200001G B42000010G
100581-746EA 1743.08 USD
100581-746 100581-744
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite

Used as a phosphorylating reagent.

Formula: C₁₂H₂₂N₃O₂P
MW: 271.301 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00797597
CAS Number: 102690-88-0

Order Now

Specification Test Results

Appearance Clear Colourless to Light Yellow Oil
Storage −20 °C Freezer, under inert atmosphere
Solubility Soluble in Chloroform, Dichloromethane, Ethyl Acetate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR