2,3-Naphthalenedialdehyde
Supplier: TORONTO RESEARCH CHEMICALS MS
Synonyms:
Naphthalene-2,3-dicarboxaldehyde
, Naphthalene-2,3-dicarbaldehyde
, NDA
N356000100MG
100571-372EA
256.92
USD
100571-372
2,3-Naphthalenedialdehyde
2,3-Naphthalenedialdehyde (NDA)
Naphthalene-2,3-dicarboxaldehyde is a fluorogenic derivatizing agent for amines.
- Synonyms: 2,3-Naphthalenedicarboxaldehyde; 2,3-Diformylnaphthalene; NSC 72106; Naphthalene-2,3-dicarbaldehyde
Formula:
C₁₂H₈O₂ MW: 184.19 g/mol Melting Pt: 128…131 °C |
MDL Number:
MFCD00016618 CAS Number: 7149-49-7 |
Specification Test Results
Appearance | Yellow Solid |
Storage | −20 °C Freezer |
Solubility | Slightly soluble in Chloroform and Ethyl Acetate |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...