WHEATON® Filtering Flask
Supplier: DWK Life Sciences
635527
26316-558CS 394.75 USD
26316-558
WHEATON® Filtering Flask
Flasks Filter Flasks
Filtering flask is used for the filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents.
  • Recommended for general filtration
  • Units have a #2 hose connection for 1/4# (6mm) I.D. flexible tubing
  • Ground glass connection eliminates phthalate contamination from silicone stopper
Certifications: Glass components are low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirement
Delivery information: Includes a coarse porosity (40-60µm) fritted glass support base, graduated funnel, anodized aluminum clamp and 1 L filter flask