(+)-Biotin-4-amidobenzoic acid, sodium salt
Supplier: TORONTO RESEARCH CHEMICALS MS
Synonyms:
BIOTIN 4-AMIDOBENZOIC ACID SODIUM SALT
B389060100MG
100582-022EA
427.69
USD
100582-022
(+)-Biotin-4-amidobenzoic acid, sodium salt
(+)-Biotin-4-amidobenzoic acid, sodium salt
(+)-Biotin 4-amidobenzoic acid, sodium salt is a fluorigenic substrate for the determination of biotinidase activity.
Formula:
C₁₇H₂₀N₃NaO₄S MW: 385.41 g/mol Melting Pt: >250 °C (dec.) |
MDL Number:
MFCD00063374 CAS Number: 102418-74-6 |
Specification Test Results
Chemical name | (+)-Biotin 4-Amidobenzoic Acid, Sodium Salt |
Synonyms | 4-[[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]benzoic Acid Sodium Salt; N-Biotinyl-p-aminobenzoic Acid Sodium Salt; |
Appearance | Off-White to Pale Yellow Solid |
Storage | Hygroscopic, -20 °C Freezer, Under Inert Atmosphere |
Solubility | Aqueous Basic, DMSO |
Stability | No Data Available |
Category | Biotin Derivatives |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...