1,4-Butanediylbismethanethiosulfonate (MTS-4-MTS)

Supplier: TORONTO RESEARCH CHEMICALS MS

Synonyms: 1,4-BUTANEDIYL BISMETHANETHIOsulfONATE[4-(mesylthio)butylthio]sulphonylmethane1,4-BUTANEDIYL BISMETHANETHIOSULPHONATE[4-(mesylthio)butylthio]sulfonylmethane1,4-bis(methylsulfonylsulfanyl)butane1,4-bis(methylsulphonylsulphanyl)butane

B69015025MG
100580-812EA 327.69 USD
100580-812
1,4-Butanediylbismethanethiosulfonate (MTS-4-MTS)
1,4-Butanediylbismethanethiosulfonate (MTS-4-MTS)

It is a MTS and sulfhydryl active reagents and also act as a sulfhydryl cross linking reagents.

Formula: C₆H₁₄O₄S₄
MW: 278.414 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD01320381
CAS Number: 55-99-2

Order Now

Specification Test Results

Appearance White solid
Solubility Dichloromethane, DMF, DMSO
Stability Moisture, Temperature Sensitive

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR