Iron(III) citrate monohydrate
Supplier: Pfaltz & Bauer
Synonyms:
Iron (III) citrate tribasic monohydrate
, Ferric citrate monohydrate
, Ferric Citrate, n-Hydrate
F00740-100G
101179-890EA
324.83
USD
101179-890
Iron(III) citrate monohydrate
Iron (III) citrate monohydrate
Formula:
C₆H₅FeO₇·H₂O MW: 262.96 g/mol Boiling Pt: 310 °C Density: 1.762 Storage Temperature: Ambient |
MDL Number:
MFCD00013098 CAS Number: 2338-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...