L(+)-Cysteine hydrochloride, anhydrous ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-2-Amino-3-mercaptopropanoic acid hydrochloride
, L-Cysteine hydrochloride
, H-Cys-OH.HCl
, (R)-(+)-Cysteine hydrochloride
075781-10G
075781-100G
102542-988EA
15.38
USD
102542-988
102542-990
L(+)-Cysteine hydrochloride, anhydrous ≥95%
L(+)-Cysteine hydrochloride
Hazard: Irritant
Formula:
HSCH₂CH(NH₂)COOH·HCl MW: 157.62 g/mol |
MDL Number:
MFCD00064553 CAS Number: 52-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...