Magnetic Film Holder, International Crystal Laboratories
Supplier: International Crystal Laboratories
0009-4450
100215-668EA 168.08 USD
100215-668
Magnetic Film Holder, International Crystal Laboratories
Film Processing Supplies
Magnetic holders for KBr pellet, 13 mm microscope slide optics or thin polymer films.
Smaller samples may be mounted on a slotted opaque card (masking the spectrophotometer beam) and then inserted into the magnetic film holder. The film sample, pellet or optic is held in place by a magnetized rubber plate. These holders fit in the universal 2×3" slide mount in all spectrophotometers. Use of paper inserts facilitates mounting small samples and pressing KBr pellets.