Total Protein Reagent, Pointe Scientific
Supplier: Pointe Scientific
OT928-576 HT928-576 HT928-1200
10022-850EA 415.59 USD
10022-850 10023-500 10023-404
Total Protein Reagent, Pointe Scientific
Assays Protein Assays
Hitachi Total Protein Reagent is for the quantitative determination of total protein concentration in serum.
  • Quantitative determination of total protein concentration in serum
  • Intensity of the violet color is proportional to the amount of protein present
  • For in vitro diagnostic use only
Hitachi Total Protein Reagent is for the quantitative determination of total protein concentration in serum
The Hitachi 576 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are six 66 mL R1 Reagent bottles and six 30 mL R2 Reagent bottles in the HT928-576 kit.
The Hitachi 1200 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are eight 100 mL R1 Reagent bottles and four 100 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An NIST-traceable protein standard or serum calibrator control sera with known normal and abnormal total protein values.