D-Mannosamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D-Mannosamine hydrochloride

J66316-03 J66316-ME
10815-554EA 145.04 USD
10815-554 10815-552
D-Mannosamine hydrochloride 98%
D-Mannosamine hydrochloride
Formula: C6H14ClNO5
MW: 215.632 g/mol
Melting Pt: 168 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064557
CAS Number: 5505-63-5
EINECS: 226-847-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR