Ward's® AP Biology Investigations Bundle

Supplier: Ward's
470164-242KT 1099.99 USD
470164-242 470164-694
Ward's® AP Biology Investigations Bundle
Educational Materials Biology Educational Materials Scientific Inquiry Learning Activities
Buy All Your Labs At Once and Save

  • Updated literature aligned with new AP Biology curriculum
  • Procedural and concept illustrations to aid comprehension
  • Materials provided for 32 students in 8 lab groups
  • A Refill Bundle is available

All inclusive AP Biology Lab Kit and Refill bundles offer convenience and savings compared to purchasing each lab or refill separately. Purchase for a single class or share among multiple sections and decide which labs work the best for you and your students.

The Only Kits Aligned with AP Science Practices. Ward's AP Biology Investigations provide complete coverage of the Four Big Ideas and Seven Science Practices to help you meet new AP Biology curriculum standards and prepare students for real-world success in college and research-level labs. Each investigation follows a series of three parts, beginning with a structured lab investigation, followed by a related guided inquiry investigation with thoughtful assessment questions, and finally, suggestions to encourage students to formulate their own questions and explore further self-guided experimentation. This complete series of investigations emphasizes the progression to fully student-directed investigations and builds competence in all of the science practices, helping students:
•Practice concept application instead of fact memorization.
•Focus on self-guided experimentation instead of step-by-step lab procedure.
•Investigate real-world concepts with greater curriculum flexibility.
•Apply problem-solving skills to strengthen student inquiry.

Our AP Biology investigation bundle includes all of our investigation kits listed below:
AP Biology Investigation 1: Evolution: Artificial Selection
AP Biology Investigation 2: Evolution: Hardy-Weinberg
AP Biology Investigation 4: Cell Processes: Diffusion and Osmosis
AP Biology Investigation 5: Cell Processes: Photosynthesis
AP Biology Investigation 6: Cell Processes: Cellular Respiration
AP Biology Investigation 7: Genetics and Information Transfer: Mitosis and Meiosis
AP Biology Investigation 8: Genetics and Information Transfer: Transformation
AP Biology Investigation 9: Genetics and Information Transfer: Restriction Enzymes
AP Biology Investigation 10: Interactions: Energy Dynamics
AP Biology Investigation 11: Interactions: Transpiration
AP Biology Investigation 12: Interactions: Behavior
AP Biology Investigation 13: Interactions: Enzyme Activity
Note- AP Biology Investigation 3: Evolution: Comparing DNA Sequences to Understand Evolutionary Relationships with BLAST is available for free online at: http://blast.ncbi.nlm.nih.gov/Blast.cgi

Our AP Investigation refill kit bundle includes the following refill kits:
AP Biology Investigation 1 Refill: Artificial Selection
AP Biology Investigation 4 Refill: Diffusion and Osmosis
AP Biology Investigation 5 Refill: Photosynthesis
AP Biology Investigation 6 Refill: Cellular Respiration
AP Biology Investigation 7 Refill: Mitosis and Meiosis
AP Biology Investigation 9 Refill: Restriction Enzymes
AP Biology Investigation 11 Refill: Transpiration
AP Biology Investigation 12 Refill: Animal Behavior
AP Biology Investigation 13 Refill: Enzyme Activity
Note: Refill kits contain only consumable component items and do not contain additional copy masters.

Accessories information: Refill kits contain only consumable component items and do not contain additional copy masters.

Ordering information: At times, supply may be limited due to high demand.  Please place your order approximately two weeks in advance.

Note: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR