Anti-CTB Goat Polyclonal Antibody

Supplier: List Biological Laboratories
703
102946-502EA 393.85 USD
102946-502
Anti-CTB Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR