Potassium pyroantimonate ≥94%
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Potassium hexahydroxoantimonate
, Potassium antimonate
, Potassium hexahydroxoantimonate(V)
20220
100504-242EA
131.3
USD
100504-242
Potassium pyroantimonate ≥94%
Potassium pyroantimonate
Ultrastructural localization sodium. Reference: J. Histochem Cystochem., 24, 740 (1976).
Formula:
KSbO₃·3H₂O MW: 262.9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011415 CAS Number: 12208-13-8 UN: 1549 ADR: 6.1,III |
Specification Test Results
Insoluble matters | Not more than 0.01% |
Min. assay | 94% |
Min. sensitivity to Na | 1:430 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...