Potassium permanganate ACS

Supplier: Electron Microscopy Sciences
Warning

20200
100504-240EA 72.6 USD
100504-240
Potassium permanganate ACS
Potassium permanganate

Potassium permanganate is an EM fixative and a metal stain. See J. Ultrastruct. Res., 21,424 (1968) for details.

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: >240 °C (>464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1,II
https://us.vwr.com/stibo/web/std.lang.all/48/86/28984886.jpg

Order Now

Specification Test Results

Chloride and chlorate 0.005%
Nitrogen compounds 0.005%
Sulfate 0.02%
Insoluble matters 0.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR