HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥97.0%

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Hexamethyldisilazane

16700
100504-866EA 75.72 USD
100504-866
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥97.0%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)

For fast preparation of soft insect tissues for SEM. A good substitution for critical point drying.

Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126 °C (259 °F)
Melting Pt: –82 °C (–116 °F)
Density: 0.774 g/cm³ (6.459 lbs/gal.)
Flash Pt: 6 °C (43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/48/32/28984832.jpg

Order Now

Specification Test Results

Assay 97.0%
Refractive Index at 20 °C 1.407
Specific Gravity at 25 °C 0.774

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR