Econo™ Sterile Adson Brown Tissue Forceps, Floor Grade, Sklar
Supplier: Sklar
96-2558 96-2559
89023-862CS 540.5 USD
89023-862 89023-864
Econo™ Sterile Adson Brown Tissue Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Adson-Brown Tissue forceps are for holding, and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7 x 7 or 9 x 9. They are used to grasp delicate tissue, in plastic surgery.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments reduce the likelihood of cross-contamination.