Potassium permanganate ≥98%

Supplier: BeanTown Chemical

Synonyms: Permanganic acid potassium salt

144570-500G
BT144570-500GEA 64.84 USD
BT144570-500G
Potassium permanganate ≥98%
Potassium permanganate

  • RTECS: SD6475000
  • Light Sensitive
  • Crystalline

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: 240 °C
Density: 2.71 g/mL
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR