HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hexamethyldisilazane
130970-500G
130970-25G
130970-100G
BT130970-500GEA
156.36
USD
BT130970-500G
BT130970-25G
BT130970-100G
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥99%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
- RTECS: JM9230000
- Refractive index: 1.408
- Moisture Sensitive
- Liquid
Formula:
(CH₃)₃SiNHSi(CH₃)₃ MW: 161.39 g/mol Boiling Pt: 125 °C Density: 0.774 g/mL Flash Pt: 11 °C (52 °F) |
MDL Number:
MFCD00008259 CAS Number: 999-97-3 EINECS: 213-668-5 UN: 2924 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...