10-Methylphenothiazine ≥98.0% (by GC)

Supplier: TCI America

M2517-5G
TCM2517-5GEA 52.81 USD
TCM2517-5G
10-Methylphenothiazine ≥98.0% (by GC)
10-Methylphenothiazine

CAS Number: 1207-72-3
MDL Number: MFCD00041836
Molecular Formula: C13H11NS
Molecular Weight: 213.30
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 102
Lambda max.: 310 nm (CHCl3)

Formula: C₁₃H₁₁NS
MW: 213.3 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00041836
CAS Number: 1207-72-3
https://us.vwr.com/stibo/web/std.lang.all/85/49/28988549.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR