2-Fluorophenethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(2-Fluorophenyl)ethylamine
, 2-Fluorophenethylamine
, 1-Amino-2-(2-fluorophenyl)ethane
CAS Number: 52721-69-4
MDL Number: MFCD00075502
Molecular Formula: C8H10FN
Molecular Weight: 139.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 77
Specific Gravity (20/20): 1.07
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Amino-2-(2-fluorophenyl)ethane, 2-Fluorophenethylamine
Formula:
C₈H₁₀FN MW: 139.17 g/mol Boiling Pt: 64 °C Density: 1.07 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00075502 CAS Number: 52721-69-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...