Anti-CD49d Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: R1-2]

Supplier: Biolegend
103605 103606
103605-BLEA 97.66 USD
103605-BL 103606-BL
Anti-CD49d Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: R1-2]
Antibodies
FITC anti-mouse CD49d [R1-2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: R1-2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR