Ethyl-4-iodobenzoate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 4-iodobenzoate
, 4-Iodobenzoic acid ethyl ester
212085-10G
212085-50G
212085-250G
BT212085-50GEA
210.59
USD
BT212085-50G
BT212085-10G
BT212085-250G
Ethyl-4-iodobenzoate ≥98%
Ethyl 4-iodobenzoate
- Refractive index: 1.588
- Light Sensitive
- Liquid
Formula:
C₉H₉IO₂ MW: 276.07 g/mol Density: 1.641 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00017344 CAS Number: 51934-41-9 EINECS: 429-550-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...