Anti-IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MOPC-21]

Supplier: Enzo Life Sciences
SAB-600FI-050
10049-468EA 280 USD
10049-468
Anti-IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MOPC-21]
Antibodies
Isotype matched controls are useful in flow cytometry to determine a gross estimate of non-specific binding that may be observed with a specific antibody of interest. In general, an isotype control should be of the same species and isotype as the antibody of interest, used at the same concentration, and conjugated to the same fluorophore. This reagent is an IgG1 isotype mouse monoclonal antibody (Clone # MOPC-21), conjugated to FITC. It is intended for use as an isotype control antibody for flow cytometry.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR