Lucifer yellow CH dilithium salt, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulphonic acid dilithium saltLY CH lithium salt6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulfonic acid dilithium salt

0215526710 0215526725 0215526780
IC15526710EA 203.29 USD
IC15526710 IC15526725 IC15526780
Lucifer yellow CH dilithium salt, powder
Lucifer yellow CH dilithium salt

Lucifer Yellow CH is the hydrazine (-NH-CO-NH-NH2) form of the dye compared to Lucifer Yellow VS, which is the vinyl sulfone (-Ph-SO2-CH=CH2) form. It is a highly fluorescent dye useful in marking nerve cells.


Lucifer yellow CH has been used to demonstrate electrical coupling between hamster oocytes and cumulous cells during meiotic maturation in vivo and in vitro. This product can be used in staining nerve cells.


Emission = 540 +/-5 nm(Lit.)


Soluble in water (1 mg/mL - clear, yellow to brownish solution); Insoluble in ethanol.


Lucifer Yellow CH is the hydrazine (-NH-CO-NH-NH2) form of the dye compared to Lucifer Yellow VS, which is the vinyl sulfone (-Ph-SO2-CH=CH2) form. It is a highly fluorescent dye useful in marking nerve cells.

Formula: C₁₃H₉N₅O₉S₂Li₂
MW: 457.25 g/mol
MDL Number: MFCD00006922
CAS Number: 67769-47-5

Order Now

Specification Test Results

Form Orange Powder
Storage Store at +4 °C, desiccate, store under nitrogen.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR