DMP (Dimethyl pimelimidate dihydrochloride), white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
DMP, Pimelimidic acid dimethyl ester dihydrochloride, Dimethyl pimelimidate dihydrochloride, Dimethyl pimelinediimidate dihydrochloride, Dimethyl heptanebis(imidate) dihydrochloride
Dimethyl Pimelimidate Dihydrochloride is a bifunctional cross linking reagent used in peptide research.
Dimethyl pimelimidate dihydrochloride is a homobifunctional cross-linking reagent used in crosslinking, protein interaction and protein modification. Dimethyl pimelimidate dihydrochloride has been used to develop a technique for isolating Enterocytozoon bieneusi spores directly from human stool samples with a high degree of purity, which opens up new approaches for studying this parasite.
Soluble in water (10 mg/ml - clear) and methanol
Caution: Research Use Only (RUO).
Formula:
C₉H₁₈N₂O₂ Melting Pt: 122 °C (dec., lit.) Storage Temperature: Freezer |
MDL Number:
MFCD00038942 CAS Number: 58537-94-3 |
|
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...