Anti-PENK Goat Polyclonal Antibody

Supplier: Prosci
46-887
10113-192EA 723 USD
10113-192
Anti-PENK Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40 kDa band in rat brain lysates after 0.2 ?g/mL antibody staining. Please note that currently we cannot find an explanation in the literatu

ProenkepH alin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PENK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR