Anti-ABCD9 Goat Polyclonal Antibody

Supplier: Prosci
46-796
10117-230EA 723 USD
10117-230
Anti-ABCD9 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 70 kDa band observed in lysates of cell lines Jurkat and MOLT4 (calculated MW of 68.6 kDa according to NP_005041.1). Recommended concentration: 0.3-1 ?g/mL.

ABCD4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR