5-(N-Ethyl-N-isopropyl)amiloride, yellow solid

Supplier: TORONTO RESEARCH CHEMICALS MS

Synonyms: EIPA

E92150025MG
100576-400EA 264.62 USD
100576-400
5-(N-Ethyl-N-isopropyl)amiloride, yellow solid
5-(N-Ethyl-N-isopropyl)amiloride
Formula: C₁₁H₁₈ClN₇O
MW: 299,763 g/mol
MDL Number: MFCD00151710
CAS Number: 1154-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR