Piperidine, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexahydropyridine
, Hexazane
, Pentamethyleneimine
1.09724.0500
EM1.09724.0500CS
2316
USD
EM1.09724.0500
Piperidine, EMSURE® for analysis, Supelco®
Piperidine
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 107.3 °C (1013 hPa) Melting Pt: –10 °C Density: 0.8622 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005979 CAS Number: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8 (3), Merck Index: 12,07621 |
Specification Test Results
Solubility | (20 °C) miscible in all proportions |
Melting Point | -10 °C |
Molar Mass | 85.15 g/mol |
Boiling Point | 106 °C (1013 hPa) |
Vapor Pressure | 34 hPa (20 °C) |
Flash Point | 16 °C |
Explosion Limit | 1.5 - 10.3 %(V) |
Density | 0.86 g/cm3 (20 °C) |
pH | 12.6 (100 g/l, H2O, 20 °C) |
Ignition Point | 320 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...