Potassium pyroantimonate ≥99.0%, crystals, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Potassium hexahydroxoantimonatePotassium antimonatePotassium hexahydroxoantimonate(V)

1.05110.0100 1.05110.0100
EM1.05110.0100CS 15416.01 USD
EM1.05110.0100 CA1.05110.0100
Potassium pyroantimonate ≥99.0%, crystals, EMSURE® for analysis, Supelco®
Potassium pyroantimonate
Formula: KSbO₃·3H₂O
MW: 262.9 g/mol
MDL Number: MFCD00011415
CAS Number: 12208-13-8
EINECS: 235-387-7
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Identity passes test
Assay (iodometric, calculated on calcined substance) ≥ 99.0 %
Free alkali (as KOH) ≤ 0.1 %
Na (Sodium) ≤ 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR