Eriochrome cyanine R for analysis, reagent for aluminium (C.I. 43820), Supelco®

Supplier: MilliporeSigma

Synonyms: Chromoxane Cyanine R

1.03164.0025 1.03164.0025
EM1.03164.0025CS 3342.35 USD
CA1.03164.0025 EM1.03164.0025
Eriochrome cyanine R for analysis, reagent for aluminium (C.I. 43820), Supelco®
Eriochrome cyanine R
Formula: C₂₃H₁₅Na₃O₉S
MDL Number: MFCD00012411
CAS Number: 3564-18-9
EINECS: 222-641-7

Order Now

Specification Test Results

Solubility 420 g/l (20 °C)
Molar Mass 536.4 g/mol
Bulk Density 590 kg/m3
pH 9.1 (10 g/l, H2O, 24 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR