Anti-FAS Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-038-C100
89155-272EA 576 USD
89155-272
Anti-FAS Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG3

Recognizes human Fas.

Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR