Anti-TNFA Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-199-C100
89153-846EA 406 USD
89153-846
Anti-TNFA Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry

Recognizes human TNF-alpha. Does not cross-react with mouse TNF-alpha or human or mouse TNF-beta.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR