Anti-S Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-163-C100
89153-786EA 547 USD
89153-786
Anti-S Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, interacting with phosphoserine surrounded by positive or neutral aa (e.g. substrates of PKA, PKB, PKC, PKG etc.).Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence.

Reconstitute with 1ml water (15 minutes at room temperature).

Ordering information: Positive control included.

Type: Primary
Antigen: S
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR