Anti-CCR8 Goat Polyclonal Antibody

Supplier: Enzo Life Sciences
210-762-C200
89146-046EA 514 USD
89146-046
Anti-CCR8 Goat Polyclonal Antibody
Antibodies
Host: Goat

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human CCR8.

Type: Primary
Antigen: CCR8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR