Anti-GST Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-618-100 905-618
200061-460EA 450 USD
CA200061-460 200061-460
Anti-GST Mouse Monoclonal Antibody
Antibodies
The GST superfamily of enzymes play an important role in detoxification of xenobiotics by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. There are four main GST classes; alpha, mu, pi, and tau. GST-P is a member of the pi class of GSTs, and its expression is specifically increased in the early stages of rat hepatocarcinogenesis. Due to its high affinity for glutathione, GST is frequently utilized as an approximately 26 kDa fusion tag for the expression and purification of recombinant proteins.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Species independent

Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR