2-Chloro-4-iodopyridine ≥98.0%
Supplier: TCI America
C2089-5G
TCC2089-5GEA
36.2
USD
TCC2089-5G
2-Chloro-4-iodopyridine ≥98.0%
2-Chloro-4-iodopyridine
CAS Number: 153034-86-7
MDL Number: MFCD01861983
Molecular Formula: C5H3ClIN
Molecular Weight: 239.44
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 110
Storage Temperature: 0-10°C
Formula:
C₅H₃ClIN MW: 239.44 g/mol Melting Pt: 44 °C Flash Pt: 110 °C |
MDL Number:
MFCD01861983 CAS Number: 153034-86-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...